Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Dinitrobiphenyl, 97%

$42.80 - $340.53

Chemical Identifiers

Chemical Name or Material 2,2'-Dinitrobiphenyl
CAS 2436-96-6
Molecular Formula C12H8N2O4
Formula Weight 244.21
MDL Number MFCD00007127
Beilstein 1579851
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1406106

 
alfa aesar™
A1406106
5g Each for $42.80

AAA1406114

 
alfa aesar™
A1406114*
25g Each for $113.91

AAA1406122

 
alfa aesar™
A1406122
100g Each for $340.53
Description & Specifications

Specifications

Formula Weight 244.21
Chemical Name or Material 2,2'-Dinitrobiphenyl
Melting Point 124° to 127°C
Boiling Point 305°C
Quantity 5g
Assay 97%
Molecular Formula C12H8N2O4
MDL Number MFCD00007127
CAS 2436-96-6
Beilstein 1579851
Solubility Information Insoluble in water
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2,2′-Dinitrobiphenyl has been used as an internal standard in the determination of p,p′-DDT in technical and formulated products using gas chromatographic method

Solubility
Insoluble in water

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.