Please login to your online account to display your discounted pricing

2,2-Dimethylglutaric Anhydride 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  116250250

 View more versions of this product

Catalog No. AC116250250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Assay ≥97.5% (titration)
Formula Weight 142.15
Quantity 25g
Color White
Beilstein 17, 418
TSCA TSCA
EINECS Number 220-922-9
Melting Point 35° to 39°C
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 MOISTURE SENSITIVE
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
MDL Number MFCD00006682
Chemical Name or Material 2, 2-Dimethylglutaric anhydride
Assay Percent Range 98%
CAS 2938-48-9
Molecular Formula C7H10O3

Specifications

Chemical Name or Material 2, 2-Dimethylglutaric anhydride
Name Note 98%
CAS 2938-48-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (titration)
Assay Percent Range 98%
Molecular Formula C7H10O3
Formula Weight 142.15
MDL Number MFCD00006682
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents