Please login to your online account to display your discounted pricing

2,2-Dimethylglutaric Anhydride 98%, ACROS Organics™

$282.99 - $282.99

Chemical Identifiers

Chemical Name or Material 2, 2-Dimethylglutaric anhydride
Name Note 98%
CAS 2938-48-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (titration)
Assay Percent Range 98%
Molecular Formula C7H10O3
Formula Weight 142.15
MDL Number MFCD00006682
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC116250250

  SDS
thermo fisher scientific
116250250
Glass bottle 25g Each for $282.99
Description & Specifications

Specifications

Physical Form Crystals or Crystalline Powder
Assay ≥97.5% (titration)
Formula Weight 142.15
Quantity 25g
Color White
Beilstein 17, 418
EINECS Number 220-922-9
TSCA TSCA
Melting Point 35° to 39°C
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 2-Dimethylglutaric anhydride
Name Note 98%
MDL Number MFCD00006682
Assay Percent Range 98%
Molecular Formula C7H10O3
CAS 2938-48-9
Safety and Handling

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.