Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethylglutaric acid, 98+%

$74.58 - $74.58

Chemical Identifiers

Chemical Name or Material 2,2-Dimethylglutaric acid
CAS 681-57-2
Molecular Formula C7H12O4
Formula Weight 160.17
MDL Number MFCD00004197
Beilstein 1768501
Synonym 2,2-Dimethylpentanedioc acid; 1,1-O-Dimethylpropane-1,3-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2048114

  SDS
alfa aesar™
B2048114
25g Each for $74.58
Description & Specifications

Specifications

Formula Weight 160.17
Melting Point 83° to 85°C
Chemical Name or Material 2,2-Dimethylglutaric acid
Beilstein 1768501
Odor Odorless
Synonym 2,2-Dimethylpentanedioc acid; 1,1-O-Dimethylpropane-1,3-dicarboxylic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
CAS 681-57-2
MDL Number MFCD00004197
Molecular Formula C7H12O4
Quantity 25g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning

Applications
2,2-Dimethylglutaric acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It hydrolyzes with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from bases, oxidizing agents, reducing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.