Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethylcyclopropylbenzene, 99%

$260.50 - $260.50

Chemical Identifiers

Chemical Name or Material 2,2-Dimethylcyclopropylbenzene
CAS 7653-94-3
Molecular Formula C11H14
Formula Weight 146.23
MDL Number MFCD01632131
Synonym 1,1-Dimethyl-2-phenylcyclopropane; 1-Phenyl-2,2-dimethylcyclopropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2127114

 
alfa aesar™
B2127114
25g Each for $260.50
Description & Specifications

Specifications

Formula Weight 146.23
Chemical Name or Material 2,2-Dimethylcyclopropylbenzene
Assay 99%
Molecular Formula C11H14
Boiling Point 67° to 69°C (10mmHg)
MDL Number MFCD01632131
CAS 7653-94-3
Quantity 25g
Synonym 1,1-Dimethyl-2-phenylcyclopropane; 1-Phenyl-2,2-dimethylcyclopropane