missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethylbutyryl chloride, 98%

$52.58 - $467.26

Chemical Identifiers

CAS 5856-77-9
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD03093071
InChI Key LDJUYMIFFNTKOI-UHFFFAOYSA-N
Synonym 2,2-dimethylbutyryl chloride, 2,2-dimethyl butyryl chloride, 2,2-dimethyl-butyryl chloride, butanoylchloride, 2,2-dimethyl, 2,2-dimethylbutyric acid chloride, butanoyl chloride, 2,2-dimethyl, 2,2-dimethylbutyrylchloride, acmc-1aznz, ksc274k4n, 2,2-dimethylbutyryl chloride 10g
PubChem CID 79958
IUPAC Name 2,2-dimethylbutanoyl chloride
SMILES CCC(C)(C)C(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1812509 View Documents Alfa Aesar™
L1812509
10g Each for $52.58
 
AAL1812518 View Documents Alfa Aesar™
L1812518
50g Each for $145.58
 
AAL1812530 View Documents Alfa Aesar™
L1812530
250g Each for $467.26
 
Description

Description

Applications
,2-Dimethylbutanoyl chloride is a reagent that is used in the synthesis of 4α,5-Dihydro Simvastatin, the drug, is sold under the trade name Zocor. A competitive inhibitor of HMG-CoA reductase. A synthetic analog of Lovastatin. Antilipemic.

Solubility
Reacts with water. Soluble in chloroform and ethyl acetate.

Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away. Protect from electrostatic charges.
Specifications

Chemical Identifiers

5856-77-9
134.603
LDJUYMIFFNTKOI-UHFFFAOYSA-N
79958
CCC(C)(C)C(=O)Cl
C6H11ClO
MFCD03093071
2,2-dimethylbutyryl chloride, 2,2-dimethyl butyryl chloride, 2,2-dimethyl-butyryl chloride, butanoylchloride, 2,2-dimethyl, 2,2-dimethylbutyric acid chloride, butanoyl chloride, 2,2-dimethyl, 2,2-dimethylbutyrylchloride, acmc-1aznz, ksc274k4n, 2,2-dimethylbutyryl chloride 10g
2,2-dimethylbutanoyl chloride

Specifications

29°C (84°F)
10g
C6H11ClO
UN2920
2,2-dimethylbutyryl chloride, 2,2-dimethyl butyryl chloride, 2,2-dimethyl-butyryl chloride, butanoylchloride, 2,2-dimethyl, 2,2-dimethylbutyric acid chloride, butanoyl chloride, 2,2-dimethyl, 2,2-dimethylbutyrylchloride, acmc-1aznz, ksc274k4n, 2,2-dimethylbutyryl chloride 10g
CCC(C)(C)C(=O)Cl
134.603
134.61
2,2-Dimethylbutyryl chloride
133°C
5856-77-9
MFCD03093071
1743935
LDJUYMIFFNTKOI-UHFFFAOYSA-N
2,2-dimethylbutanoyl chloride
79958
98%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.