missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethylbutyric acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 434861000

 View more versions of this product

Catalog No. AC434861000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 2-Dimethylbutyric acid
98.5
Colorless to Yellow
186.0°C to 187.0°C
Authentic
Glass bottle
MFCD00004200
0.928
VUAXHMVRKOTJKP-UHFFFAOYSA-N
2,2-dimethylbutanoic acid
11684
116.16
Liquid
595-37-9
100.0
0.9280g/mL
79°C
98.5% min. (GC)
C6H12O2
100 g
2,2-dimethylbutyric acid, butanoic acid, 2,2-dimethyl, 2,2-dimethyl-butanoic acid, 2,2-dimethyl butyric acid, 2,2-dimethyl butanoic acid, unii-ay606cn05o, butyric acid, 2,2-dimethyl, dimethyl ethyl acetic acid, 2,2-dimethyl-butyric acid, .alpha.,.alpha.-dimethylbutyric acid
CCC(C)(C)C(=O)O
116.16
CHEBI:38649
99%

Chemical Identifiers

595-37-9
116.16
VUAXHMVRKOTJKP-UHFFFAOYSA-N
11684
2,2-dimethylbutanoic acid
C6H12O2
MFCD00004200
2,2-dimethylbutyric acid, butanoic acid, 2,2-dimethyl, 2,2-dimethyl-butanoic acid, 2,2-dimethyl butyric acid, 2,2-dimethyl butanoic acid, unii-ay606cn05o, butyric acid, 2,2-dimethyl, dimethyl ethyl acetic acid, 2,2-dimethyl-butyric acid, .alpha.,.alpha.-dimethylbutyric acid
CHEBI:38649
CCC(C)(C)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 209-865-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only