Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethylbutane, 99%

$57.53 - $57.53

Chemical Identifiers

Chemical Name or Material 2,2-Dimethylbutane
CAS 75-83-2
Molecular Formula C6H14
Linear Formula (CH3)3CCH2CH3
Formula Weight 86.18g/mol
MDL Number MFCD00009321
UN Number UN1208
Beilstein 1730736
Synonym Neohexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL03295AE

 
alfa aesar™
L03295-AE
100mL Each for $57.53
Description & Specifications

Specifications

Melting Point −100°C
Boiling Point 49° to 50°C
Quantity 100mL
Flash Point −48°C (−54°F)
Chemical Name or Material 2,2-Dimethylbutane
CAS 75-83-2
Assay 99%
Molecular Formula C6H14
Linear Formula (CH3)3CCH2CH3
Formula Weight 86.18g/mol
MDL Number MFCD00009321
UN Number UN1208
Beilstein 1730736
Synonym Neohexane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.