missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[2-(Dimethylamino)ethoxy]benzylamine, 90%, Maybridge

Chemical Identifiers

CAS 91215-97-3
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C11H18N2O
Formula Weight 194.28
MDL Number MFCD08690290
Chemical Name or Material 2-[2-(Dimethylamino)ethoxy]benzylamine
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC51313CB SDS Fisher Scientific Company
CC51313CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC51313DA SDS Fisher Scientific Company
CC51313DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

91215-97-3
100.0
C11H18N2O
MFCD08690290
90%
89.5
90%
194.28
2-[2-(Dimethylamino)ethoxy]benzylamine

Specifications

194.28
100.0
91215-97-3
MFCD08690290
C11H18N2O
90%
250mg
Amber Glass Bottle
90%
89.5
2-[2-(Dimethylamino)ethoxy]benzylamine

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger