missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[2-(Dimethylamino)ethoxy]benzoic acid, 95%, Thermo Scientific™

Chemical Identifiers

CAS 206261-66-7
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.245
MDL Number MFCD07801180
InChI Key JZLYEZGHBQFRLU-UHFFFAOYSA-N
Synonym 2-2-dimethylamino ethoxy benzoic acid, 2-2-dimethylamino-ethoxy-benzoic acid, 2-2-dimethylaminoethoxy benzoic acid, 2-2-dimethylaminoethyloxy benzoic acid, 2-carboxy-beta-dimethylamino phenetole, 2-2-n,n-dimethylamino ethoxy benzoic acid, 2-2-n,n-dimethylamino ethyl oxy-benzoic acid
PubChem CID 6484659
IUPAC Name 2-[2-(dimethylamino)ethoxy]benzoic acid
SMILES CN(C)CCOC1=CC=CC=C1C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC51301CB
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC51301CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC51301DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC51301DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

206261-66-7
209.245
JZLYEZGHBQFRLU-UHFFFAOYSA-N
6484659
CN(C)CCOC1=CC=CC=C1C(=O)O
C11H15NO3
MFCD07801180
2-2-dimethylamino ethoxy benzoic acid, 2-2-dimethylamino-ethoxy-benzoic acid, 2-2-dimethylaminoethoxy benzoic acid, 2-2-dimethylaminoethyloxy benzoic acid, 2-carboxy-beta-dimethylamino phenetole, 2-2-n,n-dimethylamino ethoxy benzoic acid, 2-2-n,n-dimethylamino ethyl oxy-benzoic acid
2-[2-(dimethylamino)ethoxy]benzoic acid

Specifications

206261-66-7
100.0
C11H15NO3
Amber Glass Bottle
JZLYEZGHBQFRLU-UHFFFAOYSA-N
2-[2-(dimethylamino)ethoxy]benzoic acid
6484659
95%
2-[2-(Dimethylamino)ethoxy]benzoic acid
94.5
95%
MFCD07801180
2-2-dimethylamino ethoxy benzoic acid, 2-2-dimethylamino-ethoxy-benzoic acid, 2-2-dimethylaminoethoxy benzoic acid, 2-2-dimethylaminoethyloxy benzoic acid, 2-carboxy-beta-dimethylamino phenetole, 2-2-n,n-dimethylamino ethoxy benzoic acid, 2-2-n,n-dimethylamino ethyl oxy-benzoic acid
CN(C)CCOC1=CC=CC=C1C(=O)O
209.245
209.24
250mg
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'