Please login to your online account to display your discounted pricing

2-[2-(Dimethylamino)ethoxy]aniline, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC51314CB

 View more versions of this product

Catalog No. CC51314CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.25
Quantity 250mg
Assay 97%
Chemical Name or Material 2-[2-(Dimethylamino)ethoxy]aniline
Assay Percent Range 97%
Molecular Formula C10H16N2O
CAS 1202-00-2
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-[2-(Dimethylamino)ethoxy]aniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H16N2O
Formula Weight 180.25
Accessories

GHS Signal Word: Danger

SDS & Safety Documents