missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethyl-N-(2-pyridyl)propionamide, 97%

$81.73 - $311.51

Chemical Identifiers

CAS 86847-59-8
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00460277
InChI Key CGSPVYCZBDFPHJ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-n-pyridin-2-yl-propionamide, 2-pivaloylamino pyridine, 2,2-dimethyl-n-pyridin-2-yl propanamide, n-pyridin-2-yl pivalamide, n-boc-2-aminopyridine, 2,2-dimethyl-n-2-pyridyl propanamide, 2,2-dimethyl-n-2-pyridinyl propanamide, propanamide, 2,2-dimethyl-n-2-pyridinyl, pivaloylamino pyridine, n-o-pyridylpivalamide
PubChem CID 1051519
IUPAC Name 2,2-dimethyl-N-pyridin-2-ylpropanamide
SMILES CC(C)(C)C(=O)NC1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6589806 View Documents Alfa Aesar™
H6589806
5g Each for $81.73
 
AAH6589814 View Documents Alfa Aesar™
H6589814
25g Each for $311.51
 
Specifications

Chemical Identifiers

86847-59-8
178.235
CGSPVYCZBDFPHJ-UHFFFAOYSA-N
1051519
CC(C)(C)C(=O)NC1=CC=CC=N1
C10H14N2O
MFCD00460277
2,2-dimethyl-n-pyridin-2-yl-propionamide, 2-pivaloylamino pyridine, 2,2-dimethyl-n-pyridin-2-yl propanamide, n-pyridin-2-yl pivalamide, n-boc-2-aminopyridine, 2,2-dimethyl-n-2-pyridyl propanamide, 2,2-dimethyl-n-2-pyridinyl propanamide, propanamide, 2,2-dimethyl-n-2-pyridinyl, pivaloylamino pyridine, n-o-pyridylpivalamide
2,2-dimethyl-N-pyridin-2-ylpropanamide

Specifications

71°C to 75°C
86847-59-8
MFCD00460277
2,2-dimethyl-n-pyridin-2-yl-propionamide, 2-pivaloylamino pyridine, 2,2-dimethyl-n-pyridin-2-yl propanamide, n-pyridin-2-yl pivalamide, n-boc-2-aminopyridine, 2,2-dimethyl-n-2-pyridyl propanamide, 2,2-dimethyl-n-2-pyridinyl propanamide, propanamide, 2,2-dimethyl-n-2-pyridinyl, pivaloylamino pyridine, n-o-pyridylpivalamide
CC(C)(C)C(=O)NC1=CC=CC=N1
178.235
178.23
97%
5g
C10H14N2O
4178333
CGSPVYCZBDFPHJ-UHFFFAOYSA-N
2,2-dimethyl-N-pyridin-2-ylpropanamide
1051519
Solid
2,2-Dimethyl-N-(2-pyridyl)propionamide