missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6106203

 View more versions of this product

Catalog No. AAH6106203


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ticagrelor intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2,2-Dimethyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one
C10H10N2O4
1 g
KGWYDVZSGRHPFK-UHFFFAOYSA-N
2,2-dimethyl-7-nitro-4H-1,4-benzoxazin-3-one
12793543
97%
85160-83-4
MFCD11048438
2,2-dimethyl-7-nitro-2h-benzo b 1,4 oxazin-3 4h-one, 2,2-dimethyl-7-nitro-2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-7-nitro-2h-1,4-benzooxazine-3 4h-one
CC1(C)OC2=C(NC1=O)C=CC(=C2)[N+]([O-])=O
222.20
222.2

Chemical Identifiers

85160-83-4
222.20
KGWYDVZSGRHPFK-UHFFFAOYSA-N
12793543
CC1(C)OC2=C(NC1=O)C=CC(=C2)[N+]([O-])=O
C10H10N2O4
MFCD11048438
2,2-dimethyl-7-nitro-2h-benzo b 1,4 oxazin-3 4h-one, 2,2-dimethyl-7-nitro-2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-7-nitro-2h-1,4-benzooxazine-3 4h-one
2,2-dimethyl-7-nitro-4H-1,4-benzoxazin-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions