missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione 98.0+%, TCI America™
$220.65 - $749.87
Chemical Identifiers
CAS | 15231-78-4 |
---|---|
Molecular Formula | C12H12O4 |
Molecular Weight (g/mol) | 220.224 |
MDL Number | MFCD00006636 |
InChI Key | NTUAHNQWHPQAMB-UHFFFAOYSA-N |
Synonym | 1,3-dioxane-4,6-dione, 2,2-dimethyl-5-phenyl, 5-phenylmeldrum's acid, 5-phenyl-meldrum's acid, acmc-1c8jq, 2.2-dimethyl-5-phenyl-1.3-dioxane-4.6-dione, 1,3-dioxane-4,6-dione,2,2-dimethyl-5-phenyl, 2,2-dimethyl-5-phenyl-1,3 dioxane-4,6-dione |
PubChem CID | 84848 |
IUPAC Name | 2,2-dimethyl-5-phenyl-1,3-dioxane-4,6-dione |
SMILES | CC1(OC(=O)C(C(=O)O1)C2=CC=CC=C2)C |
Chemical Identifiers
15231-78-4 | |
220.224 | |
NTUAHNQWHPQAMB-UHFFFAOYSA-N | |
84848 | |
CC1(OC(=O)C(C(=O)O1)C2=CC=CC=C2)C |
C12H12O4 | |
MFCD00006636 | |
1,3-dioxane-4,6-dione, 2,2-dimethyl-5-phenyl, 5-phenylmeldrum's acid, 5-phenyl-meldrum's acid, acmc-1c8jq, 2.2-dimethyl-5-phenyl-1.3-dioxane-4.6-dione, 1,3-dioxane-4,6-dione,2,2-dimethyl-5-phenyl, 2,2-dimethyl-5-phenyl-1,3 dioxane-4,6-dione | |
2,2-dimethyl-5-phenyl-1,3-dioxane-4,6-dione |
Specifications
15231-78-4 | |
C12H12O4 | |
5 g | |
NTUAHNQWHPQAMB-UHFFFAOYSA-N | |
2,2-dimethyl-5-phenyl-1,3-dioxane-4,6-dione | |
84848 | |
≥98.0% (T) | |
2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione |
White-Yellow | |
MFCD00006636 | |
1,3-dioxane-4,6-dione, 2,2-dimethyl-5-phenyl, 5-phenylmeldrum's acid, 5-phenyl-meldrum's acid, acmc-1c8jq, 2.2-dimethyl-5-phenyl-1.3-dioxane-4.6-dione, 1,3-dioxane-4,6-dione,2,2-dimethyl-5-phenyl, 2,2-dimethyl-5-phenyl-1,3 dioxane-4,6-dione | |
CC1(OC(=O)C(C(=O)O1)C2=CC=CC=C2)C | |
220.224 | |
220.22 | |
Crystalline Powder |
Safety and Handling
TSCA : No