missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethyl-3-pentanol, 97%

Manufacturer:  Alfa Aesar™ L1154503

 View more versions of this product

Catalog No. AAL1154503



Specifications

Specifications

2,2-Dimethyl-3-pentanol
131°C to 132°C
1g
C7H16O
UN1987
2,2-dimethyl-3-pentanol, 3-pentanol, 2,2-dimethyl, 3-pentanol,2-dimethyl, 3-pentanol,2,2-dimethyl, dsstox_cid_12813, dsstox_rid_79039, dsstox_gsid_32813, hmsvxzjwpviviv-uhfffaoysa, 2,2-dimethyl-3-pentanol, 3-pentanol, 2,2-dimethyl-8ci 9ci
CCC(C(C)(C)C)O
116.204
116.2
38°C (100°F)
-5°C
3970-62-5
MFCD00004566
1731777
HMSVXZJWPVIVIV-UHFFFAOYSA-N
2,2-dimethylpentan-3-ol
19871
97%

Chemical Identifiers

3970-62-5
116.204
HMSVXZJWPVIVIV-UHFFFAOYSA-N
19871
CCC(C(C)(C)C)O
C7H16O
MFCD00004566
2,2-dimethyl-3-pentanol, 3-pentanol, 2,2-dimethyl, 3-pentanol,2-dimethyl, 3-pentanol,2,2-dimethyl, dsstox_cid_12813, dsstox_rid_79039, dsstox_gsid_32813, hmsvxzjwpviviv-uhfffaoysa, 2,2-dimethyl-3-pentanol, 3-pentanol, 2,2-dimethyl-8ci 9ci
2,2-dimethylpentan-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

Documents