Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethyl-3-pentanol, 97%

Chemical Identifiers

Chemical Name or Material 2,2-Dimethyl-3-pentanol
CAS 3970-62-5
Molecular Formula C7H16O
Formula Weight 116.2
MDL Number MFCD00004566
UN Number UN1987
Beilstein 1731777
Synonym Ethyl tert-butyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1154503

  SDS
alfa aesar™
L1154503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1154506

  SDS
alfa aesar™
L1154506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.2
Chemical Name or Material 2,2-Dimethyl-3-pentanol
MDL Number MFCD00004566
Molecular Formula C7H16O
Health Hazard 1 Warning
Synonym Ethyl tert-butyl carbinol
Beilstein 1731777
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Assay 97%
CAS 3970-62-5
UN Number UN1987
Boiling Point 131° to 132°C
Melting Point −5°C
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 38°C (100°F)
Quantity 1g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.