Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethyl-3-octanol, 98%

$22.91 - $339.03

Chemical Identifiers

Chemical Name or Material 2,2-Dimethyl-3-octanol
CAS 19841-72-6
Molecular Formula C10H22O
Formula Weight 158.29
MDL Number MFCD00021945
Synonym n-Pentyl tert-butyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2041003

  SDS
alfa aesar™
B2041003
1g Each for $22.91

AAB2041006

  SDS
alfa aesar™
B2041006
5g Each for $85.58

AAB2041014

  SDS
alfa aesar™
B2041014
25g Each for $339.03
Description & Specifications

Specifications

Formula Weight 158.29
Health Hazard 1 Warning
Chemical Name or Material 2,2-Dimethyl-3-octanol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H22O
Assay 98%
CAS 19841-72-6
Quantity 1g
MDL Number MFCD00021945
Synonym n-Pentyl tert-butyl carbinol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.