Learn More
2,2-Dimethyl-3-butenoic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6406703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,2-Dimethyl-3-butenoic acid | |
-6°C | |
0.957 g/mL | |
C6H10O2 | |
MFCD00196967 | |
UN3265 | |
3-butenoic acid, 2,2-dimethyl, 2,2-dimethyl-but-3-enoic acid, 2,2-dimethyl-3-butenoic acid, ch2=chc ch3 2cooh, 3-butenoic acid,2,2-dimethyl | |
CC(C)(C=C)C(=O)O | |
114.144 | |
114.14 | |
Liquid |
10276-09-2 | |
Colorless | |
185°C | |
1.4295 | |
1 g | |
1700758 | |
SCFWAOWWAANBPY-UHFFFAOYSA-N | |
2,2-dimethylbut-3-enoic acid | |
139122 | |
95% |
Chemical Identifiers
10276-09-2 | |
114.144 | |
SCFWAOWWAANBPY-UHFFFAOYSA-N | |
2,2-dimethylbut-3-enoic acid |
C6H10O2 | |
MFCD00196967 | |
139122 | |
CC(C)(C=C)C(=O)O |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only