missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dimethyl-3-butenoic acid, 95%

Manufacturer:  Alfa Aesar™ H64067MD

 View more versions of this product

Catalog No. AAH64067MD



Specifications

Specifications

2,2-Dimethyl-3-butenoic acid
1.4295
Colorless
250mg
C6H10O2
UN3265
3-butenoic acid, 2,2-dimethyl, 2,2-dimethyl-but-3-enoic acid, 2,2-dimethyl-3-butenoic acid, ch2=chc ch3 2cooh, 3-butenoic acid,2,2-dimethyl
CC(C)(C=C)C(=O)O
114.144
114.14
95%
0.957g/mL
185°C
-6°C
10276-09-2
MFCD00196967
1700758
SCFWAOWWAANBPY-UHFFFAOYSA-N
2,2-dimethylbut-3-enoic acid
139122
Liquid

Chemical Identifiers

10276-09-2
114.144
SCFWAOWWAANBPY-UHFFFAOYSA-N
139122
CC(C)(C=C)C(=O)O
C6H10O2
MFCD00196967
3-butenoic acid, 2,2-dimethyl, 2,2-dimethyl-but-3-enoic acid, 2,2-dimethyl-3-butenoic acid, ch2=chc ch3 2cooh, 3-butenoic acid,2,2-dimethyl
2,2-dimethylbut-3-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C

SDS Documents