Please login to your online account to display your discounted pricing

2,2-Dimethyl-3,5-hexanedione, 97+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC386320010

 View more versions of this product

Catalog No. AC386320010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.0%
Formula Weight 142.2
Quantity 1g
Color Clear colorless to pale yellow
CAS Max % 100.0
CAS Min % 97.0
Refractive Index 1.4580 to 1.4600 (20°C, 589nm)
MDL Number MFCD00142767
Chemical Name or Material 2, 2-Dimethyl-3, 5-hexanedione
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula (CH3)3CC(O)CH2C(O)CH3
Molecular Formula C8H14O2
Assay Percent Range 97+%
CAS 7307-04-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 2, 2-Dimethyl-3, 5-hexanedione
CAS 7307-04-2
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C8H14O2
Linear Formula (CH3)3CC(O)CH2C(O)CH3
Formula Weight 142.2
MDL Number MFCD00142767
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents