Please login to your online account to display your discounted pricing

2,2-Dimethyl-3-(4-methylpiperidino)propanal, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO01200CB

 View more versions of this product

Catalog No. MO01200CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 183.29
Quantity 250mg
Assay 97%
CAS Min % 97.0
Synonym 2, 2-Dimethyl-3-(4-methylpiperidino)propanal (IUPAC)
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD05022484
Chemical Name or Material 2, 2-Dimethyl-3-(4-methylpiperidino)propanal
CAS 690632-29-2
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C11H21NO
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material 2, 2-Dimethyl-3-(4-methylpiperidino)propanal
CAS 690632-29-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H21NO
Formula Weight 183.29
MDL Number MFCD05022484
Synonym 2, 2-Dimethyl-3-(4-methylpiperidino)propanal (IUPAC)
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.

SDS & Safety Documents