missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethyl-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H61025MD

 View more versions of this product

Catalog No. AAH61025MD



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2,2-Dimethyl-2H-1,4-benzoxazin-3(4H)-one
C10H11NO2
250 mg
LJRUKUQEIHJTEL-UHFFFAOYSA-N
2,2-dimethyl-4H-1,4-benzoxazin-3-one
10607395
97%
10514-70-2
MFCD11868710
2,2-dimethyl-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl, 2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2,2-dimethyl-1,4-benzoxazin-3-one, 2,2-dimethyl-4h-benzo 1,4 oxazin-3-one, 2,2-dimethyl-2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-2,3-dihydro-4h-1,4-benzooxazine-3-one
CC1(C)OC2=CC=CC=C2NC1=O
177.20
177.2

Chemical Identifiers

10514-70-2
177.20
LJRUKUQEIHJTEL-UHFFFAOYSA-N
10607395
CC1(C)OC2=CC=CC=C2NC1=O
C10H11NO2
MFCD11868710
2,2-dimethyl-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl, 2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2,2-dimethyl-1,4-benzoxazin-3-one, 2,2-dimethyl-4h-benzo 1,4 oxazin-3-one, 2,2-dimethyl-2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-2,3-dihydro-4h-1,4-benzooxazine-3-one
2,2-dimethyl-4H-1,4-benzoxazin-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions