Please login to your online account to display your discounted pricing

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC27204DA

 View more versions of this product

Catalog No. CC27204DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 176.21
Quantity 1g
Assay 97%
CAS Max % 100.0
MDL Number MFCD04115387
CAS Min % 96.5
Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carbaldehyde
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H12O2
CAS 38002-88-9

Specifications

Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carbaldehyde
CAS 38002-88-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H12O2
Formula Weight 176.21
MDL Number MFCD04115387