missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CC27204DE

 View more versions of this product

Catalog No. CC27204DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carbaldehyde
5g
96.5
97%
MFCD04115387
HGKYVFOYQUSRQN-UHFFFAOYSA-N
2,2-dimethyl-3H-1-benzofuran-7-carbaldehyde
2795471
97%
Amber Glass Bottle
38002-88-9
100.0
C11H12O2
2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carbaldehyde, 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carbaldehyde, 2,2-dimethyl-7-formyl-2,3-dihydrobenzofuran, 2,3-dihydro-2,2-dimethylbenzofuran-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carboxaldehyde, 7-benzofurancarboxaldehyde, 2,3-dihydro-2,2-dimethyl
CC1(C)CC2=CC=CC(C=O)=C2O1
176.22
176.21

Chemical Identifiers

38002-88-9
176.22
HGKYVFOYQUSRQN-UHFFFAOYSA-N
2795471
CC1(C)CC2=CC=CC(C=O)=C2O1
C11H12O2
MFCD04115387
2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carbaldehyde, 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carbaldehyde, 2,2-dimethyl-7-formyl-2,3-dihydrobenzofuran, 2,3-dihydro-2,2-dimethylbenzofuran-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carboxaldehyde, 7-benzofurancarboxaldehyde, 2,3-dihydro-2,2-dimethyl
2,2-dimethyl-3H-1-benzofuran-7-carbaldehyde
SDS Documents