Learn More
2,2-Dimethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 210510500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 2-Dimethyl-1, 3-propanediamine | |
98.5 | |
29.0°C to 31.0°C | |
0.8500g/mL | |
47°C | |
98.5% min. (GC) | |
C5H14N2 | |
H2NCH2C(CH3)2CH2NH2 | |
04,III,595 | |
2,2-dimethyl-1,3-propanediamine, neopentyldiamine, 1,3-propanediamine, 2,2-dimethyl, neopentanediamine, 1,3-diamino-2,2-dimethylpropane, 2,2-dimethyltrimethylenediamine, 1,3-propanediamine,2,2-dimethyl, neopentylenediamine, 2,3-propanediamine, acmc-209oqg | |
DDHUNHGZUHZNKB-UHFFFAOYSA-N | |
2,2-dimethylpropane-1,3-diamine | |
81770 | |
99% |
7328-91-8 | |
100.0 | |
Colorless | |
152.0°C to 154.0°C | |
Authentic | |
Glass bottle | |
1.4566 | |
50 g | |
0.85 | |
Solubility in water: miscible | |
CC(C)(CN)CN | |
102.18 | |
102.18 | |
Low Melting Solid |
Chemical Identifiers
7328-91-8 | |
102.18 | |
81770 | |
CC(C)(CN)CN |
C5H14N2 | |
DDHUNHGZUHZNKB-UHFFFAOYSA-N | |
2,2-dimethylpropane-1,3-diamine |
Safety and Handling
GHS H Statement
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 230-819-
RUO – Research Use Only