Please login to your online account to display your discounted pricing

2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, ACROS Organics™

$162.51 - $162.51

Chemical Identifiers

Chemical Name or Material 2,2-Dimethyl-1,3-dioxane-4,6-dione
Name Note 98%
CAS 2033-24-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H8O4
Formula Weight 144.13
MDL Number MFCD00006638
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC198455000

  SDS
fisher scientific acros
AC198455000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC198450250

  SDS
fisher scientific acros
198450250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC198451000

  SDS
fisher scientific acros
198451000
Glass bottle 100g Each for $162.51
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystals or Powder
Formula Weight 144.13
Color Brownish-white to beige-brown
Quantity 500g
RTECS Number JH0800000
EINECS Number 217-992-8
TSCA TSCA
Melting Point 92° to 96°C
Fieser 08,313; 10,252; 11,311; 13,172
Merck Index 15, 5885
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Water 0.1% max. (K.F.)
Synonym Malonic acid cyclic isopropylidene ester
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Warning
Chemical Name or Material 2,2-Dimethyl-1,3-dioxane-4,6-dione
Molecular Formula C6H8O4
Assay Percent Range 98%
MDL Number MFCD00006638
Name Note 98%
CAS 2033-24-1
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.