missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 187842500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 2-Dimethoxy-2-phenylacetophenone | |
24650-42-8 | |
100.0 | |
Authentic | |
Glass bottle | |
C6H5COC(OCH3)2C6H5 | |
250g | |
2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621 | |
COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
256.30 | |
256.29 |
99% | |
98.5 | |
0.2% max. (75°C, 2 hrs) | |
98.5% min. (GC) | |
C16H16O3 | |
MFCD00008475 | |
14, 3392 | |
KWVGIHKZDCUPEU-UHFFFAOYSA-N | |
2,2-dimethoxy-1,2-diphenylethanone | |
90571 | |
99% |
Safety and Handling
LIGHT SENSITIVE
EINECSNumber : 246-386-6
RTECSNumber : KM5775658
TSCA : TSCA
RUO â Research Use Only