Please login to your online account to display your discounted pricing

2,2'-Dimethoxy-1,1'-binaphthyl, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  34236-0250

 View more versions of this product

Catalog No. AC342360250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 314.38
Quantity 25g
Color White to beige
TSCA TSCA
Melting Point 198° to 202°C
CAS Max % 100.0
Synonym 1, 1'-Binaphthol dimethyl ether
Chemical Name or Material 2, 2'-Dimethoxy-1, 1'-binaphthyl
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C22H18O2
MDL Number MFCD00091146
CAS 2960-93-2
CAS Min % 97.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2, 2'-Dimethoxy-1, 1'-binaphthyl
Name Note 98%
CAS 2960-93-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C22H18O2
Formula Weight 314.38
MDL Number MFCD00091146
Synonym 1, 1'-Binaphthol dimethyl ether