Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Dimethoxy-1,1'-binaphthyl, 97%

Chemical Identifiers

Chemical Name or Material 2,2'-Dimethoxy-1,1'-binaphthyl
CAS 2960-93-2
Molecular Formula C22H18O2
Formula Weight 314.38
MDL Number MFCD00091146
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5108603

  SDS
alfa aesar™
H5108603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5108606

  SDS
alfa aesar™
H5108606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 314.38
Assay 97%
CAS 2960-93-2
Quantity 1g
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P273-P305+P351+P338-P501a
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,2'-Dimethoxy-1,1'-binaphthyl
Molecular Formula C22H18O2
MDL Number MFCD00091146
UN Number UN3077
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

Applications
2,2′-Dimethoxy-1,1′-binaphthyl is used as organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions.

Safety and Handling

Warning

GHS P Statement
P280g-P273-P305+P351+P338-P501a
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.