Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Dihydroxy-4,4'-dimethoxybenzophenone, 98%

$41.88 - $136.78

Chemical Identifiers

Chemical Name or Material 2,2'-Dihydroxy-4,4'-dimethoxybenzophenone
CAS 131-54-4
Molecular Formula C15H14O5
Formula Weight 274.27g/mol
MDL Number MFCD00009606
Beilstein 1887087
Merck Index 14,1099
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2351814

 
alfa aesar™
B23518-14
25g Each for $41.88

AAB2351822

 
alfa aesar™
B23518-22
100g Each for $136.78
Description & Specifications

Specifications

Melting Point 133° to 136°C
Quantity 25g
Chemical Name or Material 2,2'-Dihydroxy-4,4'-dimethoxybenzophenone
CAS 131-54-4
Assay 98%
Molecular Formula C15H14O5
Formula Weight 274.27g/mol
MDL Number MFCD00009606
Beilstein 1887087
Merck Index 14,1099
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DJ0900000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.