Please login to your online account to display your discounted pricing

2,2-Dihydroxy-4,4-dimethoxybenzophenone, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 2'-Dihydroxy-4, 4'-dimethoxybenzophenone
CAS 131-54-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15H14O5
Linear Formula (CH3OC6H3(OH))2CO
Formula Weight 274.26
MDL Number MFCD00009606
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC388240050

  SDS
thermo fisher scientific
AC388240050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 274.26
Quantity 5g
Color Yellow
Melting Point 131° to 135°C
Merck Index 15, 1101
CAS Min % 94.0
CAS Max % 100.0
Synonym Uvinul™ D49
CAS 131-54-4
Chemical Name or Material 2, 2'-Dihydroxy-4, 4'-dimethoxybenzophenone
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C15H14O5
MDL Number MFCD00009606
Linear Formula (CH3OC6H3(OH))2CO
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.