Please login to your online account to display your discounted pricing

2,2-Dihydroxy-4,4-dimethoxybenzophenone, 95%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC388240050

 View more versions of this product

Catalog No. AC388240050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 274.26
Quantity 5g
Color Yellow
Melting Point 131° to 135°C
Merck Index 15, 1101
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2, 2'-Dihydroxy-4, 4'-dimethoxybenzophenone
Assay Percent Range 95%
Molecular Formula C15H14O5
CAS 131-54-4
Synonym Uvinul™ D49
Infrared Spectrum Authentic
Linear Formula (CH3OC6H3(OH))2CO
MDL Number MFCD00009606
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 2'-Dihydroxy-4, 4'-dimethoxybenzophenone
CAS 131-54-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15H14O5
Linear Formula (CH3OC6H3(OH))2CO
Formula Weight 274.26
MDL Number MFCD00009606
Synonym Uvinul™ D49
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents