missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2-Difluorocyclopropylamine hydrochloride, 97%, Thermo Scientific Chemicals
$342.59 - $342.59
Chemical Identifiers
CAS | 105614-25-3 |
---|---|
Molecular Formula | C3H6ClF2N |
Molecular Weight (g/mol) | 129.535 |
MDL Number | MFCD07777150 |
InChI Key | WSSNWLHFBOKGGL-UHFFFAOYSA-N |
Synonym | 2,2-difluorocyclopropylamine hydrochloride, 2,2-difluorocyclopropan-1-amine hydrochloride, 2,2-difluorocycloprop-1-ylamine hydrochloride, 2,2-difluorocyclopropanamine hydrochloride, cyclopropanamine, 2,2-difluoro-, hydrochloride, 2,2-difluorocyclopropanamine, acmc-20aojv, 2,2-difluorocyclopropylamine, chloride, 2,2-difluorocycloprop-1-ylaminehydrochloride, 2,2-difluoro-1-cyclopropanamine hydrochloride |
PubChem CID | 20161313 |
IUPAC Name | 2,2-difluorocyclopropan-1-amine;hydrochloride |
SMILES | C1C(C1(F)F)N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH29051MC
|
Thermo Scientific Chemicals
H29051MC |
100 mg |
Each for $342.59
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105614-25-3 | |
129.535 | |
WSSNWLHFBOKGGL-UHFFFAOYSA-N | |
20161313 | |
C1C(C1(F)F)N.Cl |
C3H6ClF2N | |
MFCD07777150 | |
2,2-difluorocyclopropylamine hydrochloride, 2,2-difluorocyclopropan-1-amine hydrochloride, 2,2-difluorocycloprop-1-ylamine hydrochloride, 2,2-difluorocyclopropanamine hydrochloride, cyclopropanamine, 2,2-difluoro-, hydrochloride, 2,2-difluorocyclopropanamine, acmc-20aojv, 2,2-difluorocyclopropylamine, chloride, 2,2-difluorocycloprop-1-ylaminehydrochloride, 2,2-difluoro-1-cyclopropanamine hydrochloride | |
2,2-difluorocyclopropan-1-amine;hydrochloride |
Specifications
105614-25-3 | |
C3H6ClF2N | |
100 mg | |
2,2-difluorocyclopropylamine hydrochloride, 2,2-difluorocyclopropan-1-amine hydrochloride, 2,2-difluorocycloprop-1-ylamine hydrochloride, 2,2-difluorocyclopropanamine hydrochloride, cyclopropanamine, 2,2-difluoro-, hydrochloride, 2,2-difluorocyclopropanamine, acmc-20aojv, 2,2-difluorocyclopropylamine, chloride, 2,2-difluorocycloprop-1-ylaminehydrochloride, 2,2-difluoro-1-cyclopropanamine hydrochloride | |
C1C(C1(F)F)N.Cl | |
129.535 | |
129.54 | |
2,2-Difluorocyclopropylamine hydrochloride |
∽145°C (decomposition) | |
MFCD07777150 | |
Hygroscopic | |
WSSNWLHFBOKGGL-UHFFFAOYSA-N | |
2,2-difluorocyclopropan-1-amine;hydrochloride | |
20161313 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only