Learn More
2,2-Difluorocyclopropanemethanol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H32083MD
Description
2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.
Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Specifications
2,2-Difluorocyclopropanemethanol | |
129°C | |
MFCD06797748 | |
UN1987 | |
XOLSMTBBIZDHSG-UHFFFAOYSA-N | |
(2,2-difluorocyclopropyl)methanol | |
2761401 | |
97% |
509072-57-5 | |
C4H6F2O | |
250 mg | |
2,2-difluorocyclopropyl methanol, 2,2-difluorocyclopropanemethanol, 2,2-difluorocyclopropyl-methanol, 97+%, soqdfiarbswxuup@, 2,2-difluoro-cyclopropanemethanol, 2,2-difluorocyclopropyl-methanol, 2,2-difluoro-cyclopropyl-methanol, cyclopropanemethanol, 2,2-difluoro | |
C1C(C1(F)F)CO | |
108.088 | |
108.09 |
Chemical Identifiers
509072-57-5 | |
108.088 | |
XOLSMTBBIZDHSG-UHFFFAOYSA-N | |
2761401 | |
C1C(C1(F)F)CO |
C4H6F2O | |
MFCD06797748 | |
2,2-difluorocyclopropyl methanol, 2,2-difluorocyclopropanemethanol, 2,2-difluorocyclopropyl-methanol, 97+%, soqdfiarbswxuup@, 2,2-difluoro-cyclopropanemethanol, 2,2-difluorocyclopropyl-methanol, 2,2-difluoro-cyclopropyl-methanol, cyclopropanemethanol, 2,2-difluoro | |
(2,2-difluorocyclopropyl)methanol |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only