Please login to your online account to display your discounted pricing

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material (±)-2, 2-Difluoro-1-methylcyclopropanecarboxylic acid
Assay Percent Range 97%
Molecular Formula C5H6F2O2
Formula Weight 136.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC446630010

 
thermo fisher scientific
AC446630010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC446630050

 
thermo fisher scientific
AC446630050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.1
Quantity 1g
Chemical Name or Material (±)-2, 2-Difluoro-1-methylcyclopropanecarboxylic acid
Assay Percent Range 97%
Molecular Formula C5H6F2O2
CAS 128073-33-6