Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde, 97%

$79.58 - $601.86

Chemical Identifiers

Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde
CAS 119895-68-0
Molecular Formula C8H4F2O3
Formula Weight 186.11
MDL Number MFCD00792419
Synonym 2,2-Difluoro-4-formyl-1,3-benzodioxole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2644706

 
alfa aesar™
H2644706
5g Each for $601.86

AAH2644703

 
alfa aesar™
H2644703
1g Each for $200.11

AAH26447MD

 
alfa aesar™
H26447MD
250mg Each for $79.58
Description & Specifications

Specifications

Formula Weight 186.11
Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde
Assay 97%
Molecular Formula C8H4F2O3
MDL Number MFCD00792419
CAS 119895-68-0
Synonym 2,2-Difluoro-4-formyl-1,3-benzodioxole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.