Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Diethoxyacetophenone, 96%

$49.41 - $406.36

Chemical Identifiers

Chemical Name or Material 2,2-Diethoxyacetophenone
CAS 6175-45-7
Molecular Formula C12H16O3
Formula Weight 208.26
MDL Number MFCD00009659
Synonym Phenylglyoxal diethyl acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2446414

  SDS
alfa aesar™
B2446414
25g Each for $49.41

AAB2446422

  SDS
alfa aesar™
B2446422
100g Each for $128.38

AAB2446436

  SDS
alfa aesar™
B2446436
500g Each for $406.36
Description & Specifications

Specifications

Formula Weight 208.26
Chemical Name or Material 2,2-Diethoxyacetophenone
TSCA TSCA
Assay 96%
Molecular Formula C12H16O3
MDL Number MFCD00009659
CAS 6175-45-7
Boiling Point 131° to 134°C (10mmHg)
Flash Point >110°C (230°F)
Synonym Phenylglyoxal diethyl acetal
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g

Applications
It is an effective photoinitiator for UV-curable acrylate-based coatings, adhesives and inks. used in UV-curable system. Particularly useful for photo curing clear acrylate-based formulations where non-yellowing is important. It is also recommended for pigmented inks where shelf life and color stability are essential.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.