Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Dichlorobenzilic acid, 98+%

$71.89 - $266.30

Chemical Identifiers

Chemical Name or Material 2,2'-Dichlorobenzilic acid
CAS 3152-12-3
Molecular Formula C14H10Cl2O3
Formula Weight 297.14
MDL Number MFCD00049223
Beilstein 2387319
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0635604

 
alfa aesar™
L0635604
2g Each for $71.89

AAL0635609

 
alfa aesar™
L0635609
10g Each for $266.30
Description & Specifications

Specifications

Formula Weight 297.14
Chemical Name or Material 2,2'-Dichlorobenzilic acid
Assay ≥98%
Molecular Formula C14H10Cl2O3
MDL Number MFCD00049223
CAS 3152-12-3
Beilstein 2387319
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈162°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.