Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dichloroacetophenone, 97%

$46.32 - $318.64

Chemical Identifiers

Chemical Name or Material 2,2-Dichloroacetophenone
CAS 2648-61-5
Molecular Formula C8H6Cl2O
Formula Weight 189.04
MDL Number MFCD00000844
Beilstein 1864186
Synonym Phenacylidene chloride; alpha,alpha-Dichloroacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2347506

 
alfa aesar™
B2347506
5g Each for $46.32

AAB2347522

 
alfa aesar™
B2347522
100g Each for $318.64
Description & Specifications

Specifications

Formula Weight 189.04
Chemical Name or Material 2,2-Dichloroacetophenone
TSCA TSCA
Assay 97%
Molecular Formula C8H6Cl2O
MDL Number MFCD00000844
CAS 2648-61-5
Beilstein 1864186
Synonym Phenacylidene chloride; alpha,alpha-Dichloroacetophenone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AM7175000
Flash Point >110°C (230°F)
Boiling Point 132° to 134°C (13mmHg)
Melting Point 20° to 21°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.