missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Dichloro-3,4'-bipyridine, 95%, Thermo Scientific™

$127.85 - $452.43

Chemical Identifiers

CAS 643082-10-4
Molecular Formula C10H6Cl2N2
Molecular Weight (g/mol) 225.072
MDL Number MFCD09037431
InChI Key GLIMZUXSXNGIOM-UHFFFAOYSA-N
Synonym 2,2'-dichloro-3,4' bipyridine, 2,2'-dichloro-3,4'-bipyridine, 2,2'-dichloro-3,4'bipyridine, 2-chloro-3-2-chloropyridin-4-yl pyridine, 2-chloro-4-2-chloropyridin-3-yl pyridine, 3,4'-bi 2-chloropyridine, acmc-20aok5
PubChem CID 11299081
IUPAC Name 2-chloro-3-(2-chloropyridin-4-yl)pyridine
SMILES C1=CC(=C(N=C1)Cl)C2=CC(=NC=C2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27520MC
View Documents
Thermo Fisher Scientific
H27520MC
100mg
Each for $127.85
 
AAH27520ME
View Documents
Thermo Fisher Scientific
H27520ME
500mg
Each for $452.43
 
Description

Description

Specifications

Chemical Identifiers

643082-10-4
225.072
GLIMZUXSXNGIOM-UHFFFAOYSA-N
11299081
C1=CC(=C(N=C1)Cl)C2=CC(=NC=C2)Cl
C10H6Cl2N2
MFCD09037431
2,2'-dichloro-3,4' bipyridine, 2,2'-dichloro-3,4'-bipyridine, 2,2'-dichloro-3,4'bipyridine, 2-chloro-3-2-chloropyridin-4-yl pyridine, 2-chloro-4-2-chloropyridin-3-yl pyridine, 3,4'-bi 2-chloropyridine, acmc-20aok5
2-chloro-3-(2-chloropyridin-4-yl)pyridine

Specifications

100mg
C10H6Cl2N2
2,2'-dichloro-3,4' bipyridine, 2,2'-dichloro-3,4'-bipyridine, 2,2'-dichloro-3,4'bipyridine, 2-chloro-3-2-chloropyridin-4-yl pyridine, 2-chloro-4-2-chloropyridin-3-yl pyridine, 3,4'-bi 2-chloropyridine, acmc-20aok5
C1=CC(=C(N=C1)Cl)C2=CC(=NC=C2)Cl
225.072
225.08
2,2'-Dichloro-3,4'-bipyridine
643082-10-4
MFCD09037431
GLIMZUXSXNGIOM-UHFFFAOYSA-N
2-chloro-3-(2-chloropyridin-4-yl)pyridine
11299081
95%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications