missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Dichloro-3,3'-bipyridine, 94%

$250.91 - $250.91

Chemical Identifiers

CAS 97033-27-7
Molecular Formula C10H6Cl2N2
Formula Weight 225.08
MDL Number MFCD09037429
Chemical Name or Material 2,2'-Dichloro-3,3'-bipyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27821ME SDS Alfa Aesar™
H27821ME
500mg Each for $250.91
AAH27821MC SDS Alfa Aesar™
H27821MC
100mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97033-27-7
225.08
2,2'-Dichloro-3,3'-bipyridine
C10H6Cl2N2
MFCD09037429

Specifications

500mg
C10H6Cl2N2
MFCD09037429
2,2'-Dichloro-3,3'-bipyridine
97033-27-7
225.08
116° to 122°C
94%

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning