Please login to your online account to display your discounted pricing

Alfa Aesar™ (2,2-Dichloro-1-methylcyclopropyl)benzene, 95%

$35.86 - $139.76

Chemical Identifiers

Chemical Name or Material (2,2-Dichloro-1-methylcyclopropyl)benzene
CAS 3591-42-2
Molecular Formula C10H10Cl2
Formula Weight 201.09
MDL Number MFCD00012260
Synonym 1,1-Dichloro-2-methyl-2-phenylcyclopropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2557106

  SDS
alfa aesar™
B2557106
5g Each for $35.86

AAB2557114

  SDS
alfa aesar™
B2557114
25g Each for $139.76
Description & Specifications

Specifications

Formula Weight 201.09
Chemical Name or Material (2,2-Dichloro-1-methylcyclopropyl)benzene
TSCA TSCA
Assay 95%
Molecular Formula C10H10Cl2
MDL Number MFCD00012260
CAS 3591-42-2
Boiling Point 70° to 72°C (0.8mmHg)
Synonym 1,1-Dichloro-2-methyl-2-phenylcyclopropane
Quantity 5g