missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Dibromopropane, 95%

Manufacturer:  Alfa Aesar™ H6501406

 View more versions of this product

Catalog No. AAH6501406



Specifications

Specifications

2,2-Dibromopropane
>110°C (230°F)
113°C to 114°C
5g
C3H6Br2
MFCD00000119
2,2-dibromopropane, 2-dibrompropan, acmc-1aolk, dibromo propane, inchi=1/c3h6br2/c1-3 2,4 5/h1-2h, ksc272i9f, propane, 2,2-dibromo
CC(C)(Br)Br
201.889
201.89
95%
1.85 g/mL
1.498
Colorless to Yellow
594-16-1
(CH3)2CBr2
1731162
ARITXYXYCOZKMU-UHFFFAOYSA-N
2,2-dibromopropane
11658
Liquid

Chemical Identifiers

594-16-1
201.889
ARITXYXYCOZKMU-UHFFFAOYSA-N
11658
CC(C)(Br)Br
C3H6Br2
MFCD00000119
2,2-dibromopropane, 2-dibrompropan, acmc-1aolk, dibromo propane, inchi=1/c3h6br2/c1-3 2,4 5/h1-2h, ksc272i9f, propane, 2,2-dibromo
2,2-dibromopropane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 209-828-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents