missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-2,2'-Dibromo-1,1'-binaphthyl, 96%

$102.28 - $344.68

Chemical Identifiers

CAS 74866-28-7
Molecular Formula C20H12Br2
Molecular Weight (g/mol) 412.124
MDL Number MFCD00188005
InChI Key IJUDEFZBMMRSNM-UHFFFAOYSA-N
Synonym 1,1'-bi 2-bromonaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2,2'-dibromo 1,1' binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 2,2'-dibromo-1,1'-binaphthyl, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, racemic-2,2'-dibromo-1,1'-binaphthyl, s---2,2-dibromo-1,1'-binaphthyl
PubChem CID 619909
IUPAC Name 2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2698703
View Documents
Alfa Aesar™
H2698703
1g
Each for $102.28
 
AAH2698706
View Documents
Alfa Aesar™
H2698706
5g
Each for $344.68
 
Description

Description

Applications
The synthesis of this class of compounds relies on the lithiation of racemic 2,2′-dibromo-1,1′-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.

Solubility
Sparingly Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

74866-28-7
412.124
IJUDEFZBMMRSNM-UHFFFAOYSA-N
619909
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br
C20H12Br2
MFCD00188005
1,1'-bi 2-bromonaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2,2'-dibromo 1,1' binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 2,2'-dibromo-1,1'-binaphthyl, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, racemic-2,2'-dibromo-1,1'-binaphthyl, s---2,2-dibromo-1,1'-binaphthyl
2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene

Specifications

178°C to 183°C
74866-28-7
MFCD00188005
Sparingly Soluble in water.
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br
412.124
412.23
(+/-)-2,2'-Dibromo-1,1'-binaphthyl
1g
C20H12Br2
1,1'-bi 2-bromonaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2,2'-dibromo 1,1' binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 2,2'-dibromo-1,1'-binaphthyl, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, racemic-2,2'-dibromo-1,1'-binaphthyl, s---2,2-dibromo-1,1'-binaphthyl
IJUDEFZBMMRSNM-UHFFFAOYSA-N
2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene
619909
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS