Learn More
2,2'-Diaminodiphenyl disulfide, 97%, Thermo Scientific Chemicals
$99.04 - $415.97
Chemical Identifiers
CAS | 1141-88-4 |
---|---|
Molecular Formula | C12H12N2S2 |
Molecular Weight (g/mol) | 248.36 |
MDL Number | MFCD00007703 |
InChI Key | YYYOQURZQWIILK-UHFFFAOYSA-N |
Synonym | 2,2'-dithiodianiline, benzenamine, 2,2'-dithiobis, intramine, 2,2'-disulfanediyldianiline, 2-aminophenyl disulfide, 2,2'-diaminodiphenyldisulfide, 2,2'-diaminodiphenyl disulfide, 2-2-aminophenyl disulfanyl aniline, aniline, 2,2'-dithiodi, bis o-aminophenyl disulfide |
PubChem CID | 14358 |
IUPAC Name | 2-[(2-aminophenyl)disulfanyl]aniline |
SMILES | NC1=CC=CC=C1SSC1=CC=CC=C1N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5580706
|
Thermo Scientific Chemicals
H5580706 |
5 g |
Each for $99.04
|
|
|||||
AAH5580714
|
Thermo Scientific Chemicals
H5580714 |
25 g |
Each for $415.97
|
|
|||||
Description
2,2'-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.
Solubility
Soluble in hot methanol- almost transparent.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
1141-88-4 | |
248.36 | |
YYYOQURZQWIILK-UHFFFAOYSA-N | |
14358 | |
NC1=CC=CC=C1SSC1=CC=CC=C1N |
C12H12N2S2 | |
MFCD00007703 | |
2,2'-dithiodianiline, benzenamine, 2,2'-dithiobis, intramine, 2,2'-disulfanediyldianiline, 2-aminophenyl disulfide, 2,2'-diaminodiphenyldisulfide, 2,2'-diaminodiphenyl disulfide, 2-2-aminophenyl disulfanyl aniline, aniline, 2,2'-dithiodi, bis o-aminophenyl disulfide | |
2-[(2-aminophenyl)disulfanyl]aniline |
Specifications
1141-88-4 | |
C12H12N2S2 | |
5 g | |
2,2'-dithiodianiline, benzenamine, 2,2'-dithiobis, intramine, 2,2'-disulfanediyldianiline, 2-aminophenyl disulfide, 2,2'-diaminodiphenyldisulfide, 2,2'-diaminodiphenyl disulfide, 2-2-aminophenyl disulfanyl aniline, aniline, 2,2'-dithiodi, bis o-aminophenyl disulfide | |
YYYOQURZQWIILK-UHFFFAOYSA-N | |
2-[(2-aminophenyl)disulfanyl]aniline | |
14358 | |
97% |
91°C to 92°C | |
MFCD00007703 | |
914032 | |
Soluble in hot methanol- almost transparent. | |
NC1=CC=CC=C1SSC1=CC=CC=C1N | |
248.36 | |
248.36 | |
2,2'-Diaminodiphenyl disulfide |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 214-529-1
RTECSNumber : BX9540000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only