missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Diaminodiphenyl disulfide, 97%

Manufacturer:  Alfa Aesar™ H5580714

 View more versions of this product

Catalog No. AAH5580714



Description

Description

Applications
2,2′-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

Solubility
Soluble in hot methanol- almost transparent.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2,2'-Diaminodiphenyl disulfide
25g
C12H12N2S2
914032
Soluble in hot methanol- almost transparent.
C1=CC=C(C(=C1)N)SSC2=CC=CC=C2N
248.362
248.36
91°C to 92°C
1141-88-4
MFCD00007703
2,2'-diaminodiphenyl disulfide
YYYOQURZQWIILK-UHFFFAOYSA-N
2-[(2-aminophenyl)disulfanyl]aniline
14358
97%

Chemical Identifiers

1141-88-4
248.362
YYYOQURZQWIILK-UHFFFAOYSA-N
14358
C1=CC=C(C(=C1)N)SSC2=CC=CC=C2N
C12H12N2S2
MFCD00007703
2,2'-diaminodiphenyl disulfide
2-[(2-aminophenyl)disulfanyl]aniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 214-529-1

RTECSNumber : BX9540000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents