Learn More
2,2-Di-n-propylacetic acid, 98+%, Thermo Scientific Chemicals
$67.49 - $620.51
Chemical Identifiers
CAS | 99-66-1 |
---|---|
Molecular Formula | C8H16O2 |
Molecular Weight (g/mol) | 144.21 |
MDL Number | MFCD00002672 |
InChI Key | NIJJYAXOARWZEE-UHFFFAOYSA-N |
Synonym | valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl |
PubChem CID | 3121 |
ChEBI | CHEBI:39867 |
IUPAC Name | 2-propylpentanoic acid |
SMILES | CCCC(CCC)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1296209
|
Thermo Scientific Chemicals
A1296209 |
10 g |
Each for $67.49
|
|
|||||
AAA1296218
|
Thermo Scientific Chemicals
A1296218 |
50 g |
Each for $173.90
|
|
|||||
AAA1296230
|
Thermo Scientific Chemicals
A1296230 |
250 g |
Each for $620.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-66-1 | |
144.21 | |
NIJJYAXOARWZEE-UHFFFAOYSA-N | |
3121 | |
2-propylpentanoic acid |
C8H16O2 | |
MFCD00002672 | |
valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl | |
CHEBI:39867 | |
CCCC(CCC)C(O)=O |
Specifications
99-66-1 | |
219°C to 220°C | |
Weak, Characteristic | |
1.425 | |
10 g | |
1750447 | |
valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl | |
CCCC(CCC)C(O)=O | |
144.21 | |
CHEBI:39867 | |
≥98% |
0.905 | |
126°C (258°F) | |
C8H16O2 | |
MFCD00002672 | |
UN3265 | |
14,9913 | |
NIJJYAXOARWZEE-UHFFFAOYSA-N | |
2-propylpentanoic acid | |
3121 | |
144.21 | |
2,2-Di-n-propylacetic acid |
Safety and Handling
GHS H Statement
H360-H314-H318-H302
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c
H302-H315-H318-H360Df
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 202-777-3
RTECSNumber : YV7875000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only