Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2-Di-n-butyl-1,3-propanediol, 96%

$244.74 - $244.74

Chemical Identifiers

Chemical Name or Material 2,2-Di-n-butyl-1,3-propanediol
CAS 24765-57-9
Molecular Formula C11H24O2
Formula Weight 188.31
MDL Number MFCD01863725
Beilstein 1699746
Synonym 2-n-Butyl-2-hydroxymethyl-1-hexanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1561914

 
alfa aesar™
L1561914
25g Each for $244.74
Description & Specifications

Specifications

Formula Weight 188.31
Chemical Name or Material 2,2-Di-n-butyl-1,3-propanediol
Melting Point 38° to 42°C
Boiling Point 125° to 130°C (1mmHg)
Quantity 25g
Assay 96%
MDL Number MFCD01863725
CAS 24765-57-9
Beilstein 1699746
Synonym 2-n-Butyl-2-hydroxymethyl-1-hexanol
Molecular Formula C11H24O2

Applications
2,2-Di-n-butyl-1,3-propanediol is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.