missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Chlorophenyl)thioacetamide, 97%

$87.28 - $215.50

Chemical Identifiers

CAS 673476-96-5
Molecular Formula C8H8ClNS
Formula Weight 185.67
MDL Number MFCD00278248
Chemical Name or Material 2-(2-Chlorophenyl)thioacetamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2668503 SDS Alfa Aesar™
H2668503
1g Each for $215.50
AAH26685MD SDS Alfa Aesar™
H26685MD
250mg Each for $87.28
Specifications Description & Specifications

Chemical Identifiers

673476-96-5
185.67
2-(2-Chlorophenyl)thioacetamide
C8H8ClNS
MFCD00278248

Specifications

1g
C8H8ClNS
MFCD00278248
97%
673476-96-5
185.67
2-(2-Chlorophenyl)thioacetamide

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger