Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Chlorophenyl)benzimidazole, 98%

$69.92 - $259.16

Chemical Identifiers

Chemical Name or Material 2-(2-Chlorophenyl)benzimidazole
CAS 3574-96-7
Molecular Formula C13H9ClN2
Formula Weight 228.68
MDL Number MFCD00051632
Beilstein 173013
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0763106

  SDS
alfa aesar™
L0763106
5g Each for $69.92

AAL0763114

  SDS
alfa aesar™
L0763114
25g Each for $259.16
Description & Specifications

Specifications

Formula Weight 228.68
Chemical Name or Material 2-(2-Chlorophenyl)benzimidazole
CAS 3574-96-7
Molecular Formula C13H9ClN2
MDL Number MFCD00051632
Assay 98%
Melting Point 232° to 236°C
Beilstein 173013
Quantity 5g
RTECS Number DD6963500